Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombospondin-1 Antibody (A65M) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018348 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018348 0.025 mg
NBP105185 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018348 Supplier Novus Biologicals Supplier No. NBP1051850.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

THSD1 Monoclonal specifically detects THSD1 in Human, Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen THSD1
Applications ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone A65M
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:200, Immunohistochemistry 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide
Gene Alias thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010
Gene Symbols THSD1
Host Species Mouse
Immunogen Extracellular matrix material from cultured bovine corneal endothelial cells.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 55901
Test Specificity Highly specific for the low-calcium (0.1 mmol/L) conformation of human TSP1. There is no evidence of cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin).
Target Species Bovine, Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.