Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK gamma Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP105096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105096 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105096 Supplier Novus Biologicals Supplier No. NBP105096

Mouse Monoclonal Antibody

LDL, MDA treated Monoclonal specifically detects LDL, MDA treated in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen LDL, MDA treated
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5D8
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:8000
Gene Alias Low density lipoprotein, MDA oxidized LDL, MDA oxidized low density lipoprotein
Host Species Mouse
Immunogen Low-density lipoprotein purified from human plasma and treated with MDA and adsorbed onto aluminum hydroxide gel
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with MDA-treated LDL but not with native LDL
Target Species Non-species specific
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.