Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDNRA/Endothelin R Type A Antibody, Novus Biologicals™

Sheep Polyclonal Antibody has been used in 4 publications
$206.50 - $426.00
Specifications
Antigen | EDNRA/Endothelin R Type A |
---|---|
Concentration | 1.7 mg/mL |
Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 10-20 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 20-40 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunofluorescence, Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
EDNRA/Endothelin R Type A Polyclonal specifically detects EDNRA/Endothelin R Type A in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
EDNRA/Endothelin R Type A | |
Western Blot 1:100-1:2000, Immunohistochemistry 10-20 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 20-40 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR | |
EDNRA | |
IgG | |
Protein G purified | |
This recognizes epitopes on the cytoplasmic side of the receptor and will not block the binding of endothelins. Therefore, the cell membrane must be permeable to the. |
1.7 mg/mL | |
Immunofluorescence, Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Sheep | |
GPCR, Neuroscience, Signal Transduction | |
PBS with No Preservative | |
1909 | |
Synthetic peptide corresponding to amino acids 410-422 (QEQNHNTERSSHK) of Rat Endothelin A Receptor, conjugated to KLH. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title