Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WIF-1 Antibody (RM0144-3M51) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50
Specifications
Antigen | WIF-1 |
---|---|
Clone | RM0144-3M51 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Description
WIF-1 Monoclonal specifically detects WIF-1 in Mouse samples. It is validated for Western Blot.Specifications
WIF-1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
11197 | |
Mouse recombinant protein of WIF-1 | |
Primary | |
This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of WIF-1. |
RM0144-3M51 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
WIF-1, WNT inhibitory factor 1 | |
WIF1 | |
IgG2 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title