Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IKK gamma Inhibitor Peptide Set
SDP

Catalog No. NBP226504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226504 2 mg
NBP2265045 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP226504 Supplier Novus Biologicals™ Supplier No. NBP226504
Add to Cart
Add to Cart

Applications: In vitro assay, In vivo assay, Functional (Inhibition), Block/Neutralize

Specifications

Host Species Human, Mouse, Rat
Components 1.IKK-gamma NEMO Binding Domain (NBD) Inhibitor Peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKTALDWSWLQTE (IKK-gamma/NEMO binding sequence is underlined). Molecular weight: 3692
2.Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
For Use With (Application) Functional (Inhibition), In vitro assay, In vivo assay
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 2 mg
Molecular Weight (g/mol) 3692
Inhibitors IKK gamma Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.