Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 alpha/IL-1F1 Antibody (024), DyLight 680
SDP

Catalog No. NB073368 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073368 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073368 Supplier Novus Biologicals Supplier No. NBP290426FR

Rabbit Monoclonal Antibody

IL-1 alpha/IL-1F1 Monoclonal specifically detects IL-1 alpha/IL-1F1 in Mouse samples. It is validated for ELISA.

Specifications

Antigen IL-1 alpha/IL-1F1
Applications ELISA
Classification Monoclonal
Clone 24
Conjugate DyLight 680
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-1 alpha/IL-1F1 (NP_034684.2; Ser 115-Ser 270).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3552
Test Specificity No cross-reactivity in ELISA with: Human IL1A
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.