Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 alpha/IL-1F1 Antibody (3405), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB366772 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366772 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366772 Supplier Novus Biologicals Supplier No. FAB2001L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-1 alpha/IL-1F1 Monoclonal antibody specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF

Specifications

Antigen IL-1 alpha/IL-1F1
Applications Flow Cytometry, Neutralization, CyTOF
Classification Monoclonal
Clone 3405
Conjugate DyLight 550
Dilution Flow Cytometry, Neutralization, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-1 alpha/IL-1F1, Ser113-Ala271, Accession # P01583
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3552
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.