Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 alpha/IL-1F1 Antibody (OTI2F8), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289195 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289195 Supplier Novus Biologicals Supplier No. NBP271021APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-1 alpha/IL-1F1 Monoclonal antibody specifically detects IL-1 alpha/IL-1F1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen IL-1 alpha/IL-1F1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI2F8
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha
Host Species Mouse
Immunogen Full length human recombinant protein of human IL1A(NP_000566) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3552
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.