Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 beta/IL-1F2 Antibody (11E5), HRP, Novus Biologicals™
SDP

Catalog No. NB10073053H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073053H 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073053H Supplier Novus Biologicals Supplier No. NB10073053H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen IL-1 beta/IL-1F2
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1100000
Conjugate HRP
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Sandwich ELISA
Formulation PBS with No Preservative
Gene Alias catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbols IL1B
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant full-length human interleukin 1-beta.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Metastasis
Primary or Secondary Primary
Gene ID (Entrez) 3553
Test Specificity Does not inhibit the biological activity of IL-1.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.