Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 RAcP/IL-1 R3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176924 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176924 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176924 Supplier Novus Biologicals Supplier No. NBP176924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-1 RAcP/IL-1 R3 Polyclonal specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IL-1 RAcP/IL-1 R3
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. AF029213
Gene Alias C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta
Gene Symbols IL1RAP
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 22 amino acids near the carboxy terminus of human IL-1RAcP. The immunogen is located within the last 50 amino acids of IL-1RAcP. Amino Acid Squence: KKSPRRSSSDEQGLSYSSLKNV
Molecular Weight of Antigen 63 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3556
Test Specificity It has no cross activity to other member in the IL-1 receptor family.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.