Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 RI Antibody (JAMA-147), PerCP, Novus Biologicals™
SDP

Catalog No. NB10065865P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065865P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065865P Supplier Novus Biologicals Supplier No. NB10065865PCP
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

IL-1 RI Monoclonal specifically detects IL-1 RI in Mouse samples. It is validated for Block/Neutralize, Flow Cytometry, Immunoprecipitation.

Specifications

Antigen IL-1 RI
Applications Blocking Assay, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone JAMA-147
Conjugate PerCP
Dilution Flow Cytometry
Gene Alias CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Gene Symbols IL1R1
Host Species Armenian Hamster
Immunogen Extracellular domain of murine IL-1R.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3554
Test Specificity NB100-65865 recognizes murine CD121a, which is the 80kD type I interleukin-1 receptor. CD121a is weakly expressed on most cells and tissues including T lymphocytes, thymocytes, epithelial cells and dendritic cells, with stronger expression reported on fibroblasts. The ligands for CD121a are the interleukin-1 (IL-1) family members; IL-1 alpha; IL-1 beta and IL-1 receptor antagonist. IL-1 is a mediator of inflammation and can elicit a variety of other biological responses through binding to CD121a. Clone JAMA-147 has been reported to block IL-1 dependent cellular responses. (1).
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.