Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 RII Antibody (021), DyLight 680
SDP

Catalog No. NB079390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB079390 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB079390 Supplier Novus Biologicals Supplier No. NBP290886FR

Rabbit Monoclonal Antibody

IL-1 RII Monoclonal specifically detects IL-1 RII in Rat samples. It is validated for ELISA.

Specifications

Antigen IL-1 RII
Applications ELISA
Classification Monoclonal
Clone 21
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-1 RII (P43303; Met1-Glu355).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7850
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.