Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 RII Antibody (RM0014-7D7), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB185472549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185472549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB185472549 Supplier Novus Biologicals Supplier No. NB11085472JF549
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-1 RII Monoclonal specifically detects IL-1 RII in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IL-1RII
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0014-7D7
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Gene Symbols IL1R2
Host Species Rat
Immunogen Mouse IL-1 RII (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7850
Test Specificity This recognizes extracellular domain of mouse IL1R-II protein.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.