Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Antibody (2050B), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB295505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295505 Supplier Novus Biologicals Supplier No. FAB9210S

Rabbit Monoclonal Antibody

IL-10 Monoclonal antibody specifically detects IL-10 in Human samples. It is validated for Neutralization, Flow Cytometry

Specifications

Antigen IL-10
Applications Neutralization, Flow Cytometry
Classification Monoclonal
Clone 2050B
Conjugate Alexa Fluor 750
Dilution Neutralization, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Host Species Rabbit
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10, Met1-Asn178, Accession # P22301
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3586
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.