Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Antibody (CC320), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB10063026J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063026J 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063026J Supplier Novus Biologicals Supplier No. NB10063026APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-10 Monoclonal specifically detects IL-10 in Bovine samples. It is validated for CyTOF, ELISA, Flow Cytometry, Functional Assay.

Specifications

Antigen IL-10
Applications CyTOF, ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone CC320
Conjugate APC
Dilution Flow Cytometry
Gene Alias CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Gene Symbols IL10
Host Species Mouse
Immunogen Plasmid cDNA encoding bovine IL-10
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3586
Test Specificity Clone CC320 has been shown to neutralise the activity of bovine IL-10 as measured by the inhibition of the inhibitory activity of IL-10 on IFN gamma synthesis (2).
Target Species Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.