Learn More
Invitrogen™ IL-10 Polyclonal Antibody

Description
In sandwich ELISAs, less than 1% cross-reactivity with recombinant canine IL-10 and recombinant porcine IL-10 is observed and less than 0.4% cross-reactivity with recombinant human IL-10, recombinant mouse IL-10, recombinant rat IL-10, and recombinant feline IL-10 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.15 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | IL-10 |
| Applications | ELISA, Neutralization, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | IL10 |
| Gene Accession No. | Q28374 |
| Gene Alias | CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF |
| Gene Symbols | IL10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.