Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 R beta Antibody (MM0360-8R24), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB211654405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211654405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211654405 Supplier Novus Biologicals Supplier No. NBP211654AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-10 R beta Monoclonal specifically detects IL-10 R beta in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-10 R beta
Applications Western Blot
Classification Monoclonal
Clone MM0360-8R24
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4
Gene Symbols IL10RB
Host Species Mouse
Immunogen Human recombinant IL-10 R beta (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3588
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.