Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 R beta Rabbit anti-Mouse, Clone: 74, Novus Biologicals™
SDP

Catalog No. NB025220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025220 50 μg
NB025219 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB025220 Supplier Novus Biologicals Supplier No. NBP29073650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-10 R beta Monoclonal specifically detects IL-10 R beta in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen IL-10 R beta
Applications ELISA
Classification Monoclonal
Clone 74
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4
Host Species Rabbit
Immunogen Recombinant Mouse IL-10 R beta protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3588
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.