Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10R alpha Antibody (1B1.3a), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB392689 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392689 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392689 Supplier Novus Biologicals Supplier No. NBP308221AF488
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Mouse, Human (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen IL-10R alpha
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 1B1.3a
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Host Species Rat
Immunogen Purified recombinant extracellular region of Mouse IL-10R alpha (Uniprot: Q61727)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3587
Target Species Mouse, Human (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.