Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10R alpha Antibody (1B1.3a), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB10063577C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063577C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063577C Supplier Novus Biologicals Supplier No. NB10063577C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-10R alpha Monoclonal specifically detects IL-10R alpha in Human (Negative), Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-10R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 1B1.3a
Conjugate DyLight 650
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Gene Symbols IL10RA
Host Species Rat
Immunogen Purified recombinant extracellular region of mouse CD210
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3587.0
Test Specificity NB100-63577 recognizes mouse CDw210a, a cell surface glycoprotein of approximately 110KD, also known as Interleukin-10 receptor 1 (IL-10R1). The epitope recognised by this antibody has been localised to the ligand binding domain within the extracellular region of mouse CDw210a. Mouse CDw210a is expressed by a range of cells including thymocytes, T cells, B cells and monocytes. This clone is reported to neutralize the effects of IL-10 (1).
Target Species Human (Negative), Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.