Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10R alpha Antibody (714212), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB364381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364381 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364381 Supplier Novus Biologicals Supplier No. FAB2742M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen IL-10R alpha
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 714212
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived and S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10 R alpha , His22-Asn235, Accession # Q13651
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3587
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.