Learn More
Description
Specifications
Specifications
| Antigen | IL-10R alpha |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI1D10 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha |
| Gene Symbols | IL10RA |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
