Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10R alpha Antibody (OTI1D10), PerCP, Novus Biologicals™
SDP

Catalog No. NB289179 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289179 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289179 Supplier Novus Biologicals Supplier No. NBP271030PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen IL-10R alpha
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D10
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Host Species Mouse
Immunogen Full length human recombinant protein of human IL10RA(NP_001549) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3587
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.