Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-11 Antibody (22626), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB296833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296833 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296833 Supplier Novus Biologicals Supplier No. FAB218X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-11 Monoclonal specifically detects IL-11 in Human samples. It is validated for Neutralization, Western Blot.

Specifications

Antigen IL-11
Applications Neutralization, Western Blot
Classification Monoclonal
Clone 22626
Conjugate Alexa Fluor 532
Dilution Western Blot, Neutralization
Formulation 50mM Sodium Borate
Gene Alias Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11Oprelvekin, interleukin 11, interleukin-11
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-11, Pro22-Leu199, Accession # P20809
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3589
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.