Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-11 Antibody (22626), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB304540 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304540 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304540 Supplier Novus Biologicals Supplier No. FAB218I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-11 Monoclonal antibody specifically detects IL-11 in Human samples. It is validated for Western Blot, Neutralization

Specifications

Antigen IL-11
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 22626
Conjugate Janelia Fluor 549
Dilution Western Blot, Neutralization
Formulation 50mM Sodium Borate
Gene Alias Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11Oprelvekin, interleukin 11, interleukin-11
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-11, Pro22-Leu199, Accession # P20809
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3589
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.