Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-11 R alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB1977
This item is not returnable.
View return policy
Description
IL-11R alpha Monoclonal specifically detects IL-11R alpha in Human samples. It is validated for Western Blot.Specifications
IL-11R alpha | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q14626 | |
IL11RA | |
Mouse myeloma cell line NS0-derived recombinant human IL-11 R alpha Cys26-Val363 Accession # Q14626 | |
100 μg | |
Primary | |
Detects human IL-11 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with rmIL-11 R alpha or recombinant human IL-6 R alpha is observed. In Western blots, 25% cross-reactivity with recombinant mouse (rm) IL-11 R alpha is observed. | |
Human | |
Purified |
Western Blot | |
473143 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AI314697, GP130, IL11R alpha, IL11RA, IL-11RA, Il11ra2, NR1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3590 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction