Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-11R alpha Antibody (025), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB073041 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073041 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073041 Supplier Novus Biologicals Supplier No. NBP290397AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-11R alpha Monoclonal specifically detects IL-11R alpha in Mouse samples. It is validated for ELISA.

Specifications

Antigen IL-11R alpha
Applications ELISA
Classification Monoclonal
Clone 25
Conjugate Alexa Fluor 647
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AI314697, GP130, IL-11RA, Il11ra2, NR1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-11R alpha (NP_001156873.1; Met 1-Gln 367) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3590
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.