Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 Antibody (1-1A4) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB395422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395422 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395422 Supplier Novus Biologicals Supplier No. NBP2504400.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-12 Monoclonal antibody specifically detects IL-12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Radioimmune Assays

Specifications

Antigen IL-12
Applications Western Blot, ELISA, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 1-1A4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
Formulation PBS
Gene Alias CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Host Species Mouse
Immunogen Recombinant human IL-12 produced in Baculovirus.
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3592
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.