Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-12 Antibody (1-1A4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$208.00
Specifications
Antigen | IL-12 |
---|---|
Clone | 1-1A4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Radioimmune Assays (RIA) |
Description
IL-12 Monoclonal antibody specifically detects IL-12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Radioimmune AssaysSpecifications
IL-12 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
Unconjugated | |
Mouse | |
Immunology, Innate Immunity | |
PBS | |
3592 | |
IgG1 | |
Protein A purified |
1-1A4 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 | |
Recombinant human IL-12 produced in Baculovirus. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title