Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 Antibody (1-1D5), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309401 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309401 Supplier Novus Biologicals Supplier No. NBP250441MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-12 Monoclonal antibody specifically detects IL-12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Functional Assay, Radioimmune Assays

Specifications

Antigen IL-12
Applications Western Blot, ELISA, Immunohistochemistry, Functional Assay, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 1-1D5
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Functional, Radioimmunoassay
Formulation 50mM Sodium Borate
Gene Alias CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Host Species Mouse
Immunogen Recombinant human IL-12 produced by baculovirus.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration
Primary or Secondary Primary
Gene ID (Entrez) 3592
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.