Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12/IL-23 p40 Antibody (171), PE, Novus Biologicals™
SDP

Catalog No. NB292613 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292613 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292613 Supplier Novus Biologicals Supplier No. NBP326107PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-12/IL-23 p40 Monoclonal antibody specifically detects IL-12/IL-23 p40 in Mouse samples. It is validated for ELISA

Specifications

Antigen IL-12/IL-23 p40
Applications ELISA
Classification Monoclonal
Clone 171
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse IL-12 p40/IL12B (Uniprot#: P43432, Met1-Ser335)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3593
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.