Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12/IL-35 p35 Antibody (2038C) [DyLight 650], Novus Biologicals™
SDP

Catalog No. NB399438 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399438 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399438 Supplier Novus Biologicals Supplier No. FAB15701W
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-12/IL-35 p35 Monoclonal antibody specifically detects IL-12/IL-35 p35 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry

Specifications

Antigen IL-12/IL-35 p35
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 2038C
Conjugate DyLight 650
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias CLMF;IL12A;IL-12A;IL-12p35;Ll12a;NFSK;NKSF1;P35
Host Species Rabbit
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 p40 and human IL-12 p35, Ile23-Ser328 of p40, Arg23-Ser219 of p35, Accession # P29460 (p40) & P29459 (P35)
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.