Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12/IL-35 p35 Antibody (27537) [Biotin], Novus Biologicals™
SDP

Catalog No. NB399434 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399434 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399434 Supplier Novus Biologicals Supplier No. FAB1570B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-12/IL-35 p35 Monoclonal antibody specifically detects IL-12/IL-35 p35 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen IL-12/IL-35 p35
Applications Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 27537
Conjugate Biotin
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CLMF;IL12A;IL-12A;IL-12p35;Ll12a;NFSK;NKSF1;P35
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12/IL-35 p35, Arg23-Ser219, Accession # P29459
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.