Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-12/IL-35 p35 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
$184.44 - $532.44
Specifications
Antigen | IL-12/IL-35 p35 |
---|---|
Clone | 2038C |
Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB15701SP
|
R&D Systems
MAB15701SP |
25 μg |
Each for $184.44
|
|
|||||
MAB15701100
|
R&D Systems
MAB15701100 |
100 μg |
Each for $532.44
|
|
|||||
Description
IL-12/IL-35 p35 Monoclonal specifically detects IL-12/IL-35 p35 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry.Specifications
IL-12/IL-35 p35 | |
Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12 p40 and human IL-12 p35 Ile23-Ser328 of p40, Arg23-Ser219 of p35 Accession # P29460 (p40) & P29459 (P35) | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-12/IL-35 p35 in direct ELISAs. Detects the IL-12/IL-35 p35 subunit either as part of the IL-12 p40/p35 heterodimer or as the IL-12/IL-35 p35 monomer. This antibody does not detect the IL-12/IL-23 p40 subunit. |
2038C | |
Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P29460 (p40) & P29459 (P35) | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title