Learn More
IL-12 (p40/p70) Rat anti-Mouse, PE, Clone: C15.6, BD
Rat Monoclonal Antibody
$116.50 - $332.00
Specifications
Antigen | IL-12 (p40/p70) |
---|---|
Clone | C15.6 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The C15.6 monoclonal antibody specifically binds to both free and complexed (homodimer p80 and heterodimer p70) forms of the p40 subunit of mouse interleukin-12 (IL-12). The immunogen used to generate the C15.6 hybridoma was recombinant mouse IL-12 p70 protein. p40 has also been described as a subunit of IL-23 and thus it is possible that the C15.6 antibody will crossreact with IL-23.
Specifications
IL-12 (p40/p70) | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
CHO-expressed recombinant mouse IL-12 p70 protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
C15.6 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.