Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 p70/IL-12A Antibody (006), PE, Novus Biologicals™
SDP

Catalog No. NB308401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308401 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308401 Supplier Novus Biologicals Supplier No. NBP289206PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-12 p70/IL-12A Monoclonal antibody specifically detects IL-12 p70/IL-12A in Human samples. It is validated for ELISA

Specifications

Antigen IL-12 p70/IL-12A
Applications ELISA
Classification Monoclonal
Clone 006
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-12 p70/IL-12A (Accession#: P29459; Met 1-Ser 219).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration
Primary or Secondary Primary
Gene ID (Entrez) 3592
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.