Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 p70/IL-12A Antibody (2Y37), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP222038W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222038W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222038W Supplier Novus Biologicals Supplier No. NBP222038AF405

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IL-12 p70/IL-12A Monoclonal specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen IL-12 p70/IL-12A
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 2Y37
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry
Gene Accession No. P29459
Gene Alias CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Gene Symbols IL12A
Host Species Mouse
Immunogen Human recombinant IL-12 p70/IL-12A
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3592
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.