Learn More
Description
Specifications
Specifications
| Antigen | IL-12 p70/IL-12A |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2Y37 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, Flow Cytometry |
| Gene Accession No. | P29459 |
| Gene Alias | CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35 |
| Gene Symbols | IL12A |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
