Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 p70/IL-12A Antibody (OTI2A8), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP271031A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271031A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271031A32 Supplier Novus Biologicals Supplier No. NBP271031AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-12 p70/IL-12A Monoclonal specifically detects IL-12 p70/IL-12A in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen IL-12 p70/IL-12A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2A8
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Gene Symbols IL12A
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 57-253 of human IL-12 p70/IL-12A (NP_000873) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration
Primary or Secondary Primary
Gene ID (Entrez) 3592.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.