Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 R beta 1 Antibody (MM0363-7V24), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP211659X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211659X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211659X Supplier Novus Biologicals Supplier No. NBP211659AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-12 R beta 1 Monoclonal specifically detects IL-12 R beta 1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-12 R beta 1
Applications Flow Cytometry
Classification Monoclonal
Clone MM0363-7V24
Conjugate Alexa Fluor 488
Dilution Flow Cytometry
Gene Accession No. P42701
Gene Alias CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL-12R subunit beta-1, IL12RB, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Gene Symbols IL12RB1
Host Species Mouse
Immunogen Human recombinant IL-12 R beta 1 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3594
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.