Learn More
Description
Ensure accurate, reproducible results in ELISA
Specifications
Specifications
| Antigen | IL-13 |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 13A |
| Concentration | 0.5 mg/mL |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry, ELISA |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 |
| Gene Symbols | IL13 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
