Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 Antibody (13A), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NBP143239A Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143239A 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143239A Supplier Novus Biologicals Supplier No. NBP143239APC
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ensure accurate, reproducible results in ELISA

IL-13 Monoclonal specifically detects IL-13 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-13
Applications Flow Cytometry
Classification Monoclonal
Clone 13A
Concentration 0.5mg/mL
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
Gene Symbols IL13
Host Species Rat
Immunogen The immunogen for this antibody was IL13.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3596
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.