Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 Antibody (13A), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB358878 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358878 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358878 Supplier Novus Biologicals Supplier No. NBP143239MFV500
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-13 Monoclonal antibody specifically detects IL-13 in Mouse samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen IL-13
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 13A
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
Host Species Rat
Immunogen The immunogen for this antibody was IL13.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3596
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.