Learn More
Description
Specifications
Specifications
| Antigen | IL-13 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | A130D 12G5 1E4 |
| Conjugate | DyLight 755 |
| Dilution | Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Gene Accession No. | P35225 |
| Gene Alias | ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 |
| Gene Symbols | IL13 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
