Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 Antibody (A130D 12G5 1E4), PE, Novus Biologicals™
SDP

Catalog No. NBP152466PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152466PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152466PE Supplier Novus Biologicals Supplier No. NBP152466PE

Mouse Monoclonal Antibody

IL-13 Monoclonal specifically detects IL-13 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-13
Applications Flow Cytometry
Classification Monoclonal
Clone A130D 12G5 1E4
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. P35225
Gene Alias ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
Gene Symbols IL13
Host Species Mouse
Immunogen A recombinant rat IL-13.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3596
Test Specificity Recognizes recombinant and native rat IL-13.
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.