Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 Antibody (B-B13), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB378030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378030 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378030 Supplier Novus Biologicals Supplier No. NBP314573AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-13 Monoclonal antibody specifically detects IL-13 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen IL-13
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone B-B13
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
Host Species Mouse
Immunogen Recombinant human IL-13
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3596
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.