Learn More
Invitrogen™ IL-13 Monoclonal Antibody (eBio13A), eBioscience™, Invitrogen™

Description
Description: The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12) Applications Reported: The eBio13A antibody has been reported useful for ELISA and ELISPOT capture, as well as intracellular staining for flow cytometric analysis. Applications Tested: The eBio13A antibody has been tested as the capture antibody in a sandwich ELISA (and ELISPOT) for analysis of mouse IL-13, in combination with the biotinylated eBio1316H antibody for detection (13-7135) and recombit mouse IL-13 (14-8131) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1.0 - 4.0 μg/mL. A standard curve consisting of doubling dilutions of the recombit standard over the range of 1000 pg/mL - 8.0 pg/mL should be included in each ELISA plate. The Functional Grade Purified eBio1316H antibody (16-7135) is recommended for in vitro blocking studies. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC.
Specifications
Specifications
| Antigen | IL-13 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | eBio13A |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Il13 |
| Gene Accession No. | P20109 |
| Gene Alias | ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.