Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-13 Monoclonal Antibody (PVM13-1), Functional Grade, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

$143.00

Specifications

Antigen IL-13
Clone PVM13-1
Concentration 1 mg/mL
Content And Storage 4°C
Applications Functional Assay, Immunostaining, Neutralization
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-147-25
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
16713981
50 μg
Each of 1 for $143.00
Only null left
Add to Cart
 
Description

Description

Description: The PVM13-1 antibody reacts with human interleukin-13 (IL-13). IL-13, also known as NC30, is a 12.5 kDa protein secreted by activated T helper cells, CD8^+ T cells, and NK cells. The biological activities of IL-13 include suppression of macrophage cytotoxic activity, upregulation of IL-1RA expression and suppression of inflammatory cytokine expression. Applications Reported: The PVM13-1 antibody has been reported for use as the capture antibody in an IL-13 ELISPOT assay or ELISA, as well as for neutralization of IL-13 bioactivity. Applications Tested: The functional grade PVM13-1 antibody has been tested for ELISA and ELISPOT capture and neutralization of IL-13 bioactivity. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Specifications

Specifications

IL-13
1 mg/mL
Functional Assay, Immunostaining, Neutralization
Functional Grade
Mouse
RUO
PBS with no preservative; pH 7.2
P35225
3596
Primary
Affinity Chromatography
PVM13-1
4°C
Monoclonal
Liquid
IgG1
Human
Il13
ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Il13
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.