Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13R alpha 1 Antibody (122), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB073177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073177 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073177 Supplier Novus Biologicals Supplier No. NBP290409UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-13R alpha 1 Monoclonal specifically detects IL-13R alpha 1 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen IL-13R alpha 1
Applications ELISA
Classification Monoclonal
Clone 122
Conjugate DyLight 350
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-13R alpha 1 (NP_598751.3; Met 1-Thr 340).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3597
Test Specificity No cross-reactivity in ELISA with: Human IL13RA1/ CD213A1 Mouse IL13RA2/ CD213A2
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.