Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13R alpha 1 Antibody (419718) [Biotin], Novus Biologicals™
SDP

Catalog No. NB399896 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399896 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399896 Supplier Novus Biologicals Supplier No. FAB1462B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-13R alpha 1 Monoclonal specifically detects IL-13R alpha 1 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen IL-13R alpha 1
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 419718
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 1, Ala27-Thr343, Accession # P78552
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3597
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.